Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isobutylbenzene ≥99.0%

Supplier: Decon Laboratories Ltd

I0097-25ML I0097-500ML
TCI0097-025MLEA 31.53 USD InStock
TCI0097-025ML TCI0097-500ML
Isobutylbenzene ≥99.0%
Isobutylbenzene
CAS Number: 538-93-2
MDL Number: MFCD00008936
Molecular Formula: C10H14
Molecular Weight: 134.22
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 172
Melting point (°C): -52
Flash Point (°C): 55
Specific Gravity (20/20): 0.85


Formula: (C₆H₅)CH₂CH(CH₃)CH₃
Boiling Pt: 172 °C
Melting Pt: -52 °C
Density: 0.85 (20 °C)
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00008936
CAS Number: 538-93-2
EINECS: 208-706-2
UN: 2709
ADR: 3,III
Merck Index: 13,05151
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=855fd3b4f5065a26b250ae6d94925fa63c1a28e4&vid=855fd3b4f5065a26b250ae6d94925fa63c1a28e4&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty