Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isopropylcyclohexane ≥99.0%

Supplier: Decon Laboratories Ltd

I0161-500ML I0161-25ML
TCI0161-500MLEA 372 USD InStock
TCI0161-500ML TCI0161-025ML
Isopropylcyclohexane ≥99.0%
Isopropylcyclohexane
CAS Number: 696-29-7
MDL Number: MFCD00001480
Molecular Formula: C9H18
Molecular Weight: 126.24
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 151
Flash Point (°C): 35
Specific Gravity (20/20): 0.80

Other Product Names / Synonyms: Hexahydrocumene


Formula: C₉H₁₈
Boiling Pt: 151 °C
Melting Pt: –90 °C
Density: 0.80 (20 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00001480
CAS Number: 696-29-7
EINECS: 211-792-4
UN: 3295
ADR: 3,I
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70e10e5249caabaf28e31f066c1bdab7ee4b246b&vid=70e10e5249caabaf28e31f066c1bdab7ee4b246b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty