Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isoindoline ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

I0751-1G I0751-5G
TCI0751-1GEA 104 USD InStock
TCI0751-1G TCI0751-5G
Isoindoline ≥98.0% (by titrimetric analysis)
Isoindoline
CAS Number: 496-12-8
MDL Number: MFCD00605324
Molecular Formula: C8H9N
Molecular Weight: 119.17
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 113
Melting point (°C): 17
Flash Point (°C): 94
Specific Gravity (20/20): 1.06
Storage Temperature: <0°C


Formula: C₈H₉N
Boiling Pt: 113 °C
Melting Pt: 17 °C
Density: 1.06 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD00605324
CAS Number: 496-12-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc8b69a7bfc852c6e0eb902aa21e3eac81e34350&vid=fc8b69a7bfc852c6e0eb902aa21e3eac81e34350&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty