Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methylhexane ≥95.0%

Supplier: Decon Laboratories Ltd
Danger

M0340-25ML M0340-5ML
TCM0340-025MLEA 336 USD InStock
TCM0340-025ML TCM0340-005ML
3-Methylhexane ≥95.0%
3-Methylhexane
CAS Number: 589-34-4
MDL Number: MFCD00009408
Molecular Formula: C7H16
Molecular Weight: 100.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 91
Flash Point (°C): -1
Specific Gravity (20/20): 0.69


Formula: C₇H₁₆
Boiling Pt: 91 °C
Melting Pt: -119.4 °C
Density: 0.69 (20 °C)
Flash Pt: -1 °C
Storage Temperature: Ambient
MDL Number: MFCD00009408
CAS Number: 589-34-4
EINECS: 209-643-3
UN: 1206
ADR: 3,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50af7273333950698623c967edc5047f1d3eab66&vid=50af7273333950698623c967edc5047f1d3eab66&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty