Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitrosalicylic acid ≥98.0%

Supplier: Decon Laboratories Ltd
Warning

N0256-5G N0256-25G
TCN0256-5GEA 146 USD InStock
TCN0256-5G TCN0256-25G
4-Nitrosalicylic acid ≥98.0%
4-Nitrosalicylic acid
CAS Number: 619-19-2
MDL Number: MFCD00071505
Molecular Formula: C7H5NO5
Molecular Weight: 183.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 234

Other Product Names / Synonyms: 2-Hydroxy-4-nitrobenzoic Acid


Formula: C₇H₅NO₅
MW: 183.12 g/mol
Melting Pt: 234 °C
MDL Number: MFCD00071505
CAS Number: 619-19-2
EINECS: 210-584-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27d423f49727d1cbb75bafc6ee5bb260afafbe40&vid=27d423f49727d1cbb75bafc6ee5bb260afafbe40&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty