Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Nitro-2,3-xylidine ≥98.0%

Supplier: Decon Laboratories Ltd

N0500-25G N0500-500G
TCN0500-025GEA 64.9 USD InStock
TCN0500-025G TCN0500-500G
6-Nitro-2,3-xylidine ≥98.0%
6-Nitro-2,3-xylidine
CAS Number: 59146-96-2
MDL Number: MFCD00010148
Molecular Formula: C8H10N2O2
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 118

Other Product Names / Synonyms: 2,3-Dimethyl-6-nitroaniline


Formula: C₈H₁₀N₂O₂
Melting Pt: 118 °C
MDL Number: MFCD00010148
CAS Number: 59146-96-2
UN: 2811
ADR: 6.1,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2f92da5462a87caf2096968c0192da1776fe9b8&vid=d2f92da5462a87caf2096968c0192da1776fe9b8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty