Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Octacosanol ≥93.0%

Supplier: Decon Laboratories Ltd

O0199-1G O0199-100MG
TCO0199-001GEA 501 USD InStock
TCO0199-001G TCO0199-100MG
1-Octacosanol ≥93.0%
1-Octacosanol
CAS Number: 557-61-9
MDL Number: MFCD00044770
Molecular Formula: C28H58O
Molecular Weight: 410.77
Purity/Analysis Method: >93.0% (GC)
Form: Crystal
Melting point (°C): 83


Formula: H₃C(CH₂)₂₆CH₂OH
Boiling Pt: 428 °C (1013 hPa)
Melting Pt: 83 °C
MDL Number: MFCD00044770
CAS Number: 557-61-9
EINECS: 209-181-2
Merck Index: 12,06840
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=966d07a52f1a47bf152e6154f3962c0fd7dcbf8d&vid=966d07a52f1a47bf152e6154f3962c0fd7dcbf8d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >93.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty