Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridine-2,3-dicarbonitrile ≥98.0%

Supplier: Decon Laboratories Ltd

P1218-1G P1218-5G
TCP1218-001GEA 119 USD InStock
TCP1218-001G TCP1218-005G
Pyridine-2,3-dicarbonitrile ≥98.0%
Pyridine-2,3-dicarbonitrile
CAS Number: 17132-78-4
MDL Number: MFCD00191682
Molecular Formula: C7H3N3
Molecular Weight: 129.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 79

Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 2,3-Dicyanopyridine


Formula: C₇H₃N₃
Melting Pt: 79 °C
MDL Number: MFCD00191682
CAS Number: 17132-78-4
EINECS: 249-492-0
UN: 3439
ADR: 6.1,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eccb2a91725727455bf9bb163d53c82d12c6bc00&vid=eccb2a91725727455bf9bb163d53c82d12c6bc00&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty