Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Phenylsuccinimide ≥98.0%

Supplier: Decon Laboratories Ltd

P1359-25G P1359-5G
TCP1359-25GEA 261 USD InStock
TCP1359-25G TCP1359-005G
N-Phenylsuccinimide ≥98.0%
N-Phenylsuccinimide
CAS Number: 83-25-0
MDL Number: MFCD00022587
Molecular Formula: C10H9NO2
Molecular Weight: 175.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 155
Storage Temperature: 0-10°C


Formula: C₁₀H₉NO₂
Melting Pt: 155 °C
MDL Number: MFCD00022587
CAS Number: 83-25-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56d23c58ec08b15d0eb771ac32233a7a580103be&vid=56d23c58ec08b15d0eb771ac32233a7a580103be&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty