Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Stearoyl chloride ≥97.0%

Supplier: Decon Laboratories Ltd

S0404-25G S0404-250G
TCS0404-025GEA 73.15 USD InStock
TCS0404-025G TCS0404-250G
Stearoyl chloride ≥97.0%
Stearoyl chloride
CAS Number: 112-76-5
MDL Number: MFCD00000744
Molecular Formula: C18H35ClO
Molecular Weight: 302.93
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 215
Flash Point (°C): 165
Freezing point (°C): 20
Specific Gravity (20/20): 0.90


Formula: C₁₈H₃₅ClO
Boiling Pt: 215 °C
Melting Pt: 164…167 °C
Density: 0.90 (20 °C)
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00000744
CAS Number: 112-76-5
EINECS: 204-004-5
UN: 3265
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3ef488a86ef3293740bd077f7d2790df215b7f0&vid=a3ef488a86ef3293740bd077f7d2790df215b7f0&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty