T0200-25ML
T0200-500ML
TCT0200-025MLEA
253
USD
InStock
TCT0200-025ML
TCT0200-500ML
Ethyl thiocyanate ≥98.0%
Ethyl thiocyanate
CAS Number: 542-90-5
MDL Number: MFCD00001834
Molecular Formula: C3H5NS
Molecular Weight: 87.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 145
Flash Point (°C): 42
Specific Gravity (20/20): 1.01
Other Product Names / Synonyms: Ethyl Rhodanide, Thiocyanic Acid Ethyl Ester