Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl trichloroacetate ≥97.0%

Supplier: Decon Laboratories Ltd
Warning

T1504-25G T1504-500G
TCT1504-025GEA 46 USD InStock
TCT1504-025G TCT1504-500G
Ethyl trichloroacetate ≥97.0%
Ethyl trichloroacetate
CAS Number: 515-84-4
MDL Number: MFCD00000795
Molecular Formula: C4H5Cl3O2
Molecular Weight: 191.43
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 167
Flash Point (°C): 65
Specific Gravity (20/20): 1.39

Other Product Names / Synonyms: Trichloroacetic Acid Ethyl Ester


Formula: C₄H₅Cl₃O₂
Boiling Pt: 167 °C
Density: 1.39 (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00000795
CAS Number: 515-84-4
EINECS: 208-212-7
UN: 2810
ADR: 6.1,I
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e365fee4d160dd54a2219645415cdfe9c612124d&vid=e365fee4d160dd54a2219645415cdfe9c612124d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty