Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzoylacetanilide ≥98.0%

Supplier: Decon Laboratories Ltd

B0904-5G B0904-25G
TCB0904-005GEA 38.7 USD InStock
TCB0904-005G TCB0904-025G
2-Benzoylacetanilide ≥98.0%
2-Benzoylacetanilide
CAS Number: 959-66-0
MDL Number: MFCD00003084
Molecular Formula: C15H13NO2
Molecular Weight: 239.27
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 107


Formula: C₁₅H₁₃NO₂
Melting Pt: 107 °C
MDL Number: MFCD00003084
CAS Number: 959-66-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fdcc458326131b8fcc0ca5e47a63e247ddff9134&vid=fdcc458326131b8fcc0ca5e47a63e247ddff9134&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty