Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bansyl chloride ≥98.0% (by HPLC)

Supplier: Decon Laboratories Ltd

B0994-1G B0994-5G
TCB0994-001GEA 671.6 USD InStock
TCB0994-001G TCB0994-5G
Bansyl chloride ≥98.0% (by HPLC)
Bansyl chloride
[N-Protecting Agent for Peptide Research]
CAS Number: 43040-76-2
MDL Number: MFCD00059134
Molecular Formula: C18H24ClNO2S
Molecular Weight: 353.91
Purity/Analysis Method: >98.0% (HPLC)
Form: Clear Liquid
Storage Temperature: 0-10°C

Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: 5-(Dibutylamino)naphthalene-1-sulfonyl Chloride


Formula: C₁₈H₂₄ClNO₂S
MDL Number: MFCD00059134
CAS Number: 43040-76-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80b69fbce2b2a83f93e6b67c3de84c80ec8f9c22&vid=80b69fbce2b2a83f93e6b67c3de84c80ec8f9c22&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty