Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture) ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

B1005-25ML B1005-500ML
TCB1005-025MLEA 40.67 USD InStock
TCB1005-025ML TCB1005-500ML
1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture) ≥98.0% (by GC, titration analysis)
1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture)
CAS Number: 2579-20-6
MDL Number: MFCD00001522
Molecular Formula: C8H18N2
Molecular Weight: 142.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 113
Specific Gravity (20/20): 0.94

Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 1,3-Di(aminomethyl)cyclohexane, Hexahydro-m-xylylenediamine


Formula: C₈H₁₈N₂
Density: 0.94 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00001522
CAS Number: 2579-20-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d13dff22e37d77ef02b39a760205acd4ac346f92&vid=d13dff22e37d77ef02b39a760205acd4ac346f92&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty