Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(4-nitrophenyl)phosphate ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

B1098-1G B1098-10G
TCB1098-001GEA 299 USD InStock
TCB1098-001G TCB1098-010G
Bis(4-nitrophenyl)phosphate ≥98.0% (by titrimetric analysis)
Bis(4-nitrophenyl)phosphate
[for Phosphodiesterase Substrate]
CAS Number: 645-15-8
MDL Number: MFCD00007318
Molecular Formula: C12H9N2O8P
Molecular Weight: 340.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 176

Other Product Names / Synonyms: Phosphoric Acid Bis(4-nitrophenyl) Ester


Formula: C₁₂H₉N₂O₈P
Melting Pt: 176 °C
MDL Number: MFCD00007318
CAS Number: 645-15-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d779d50cfe3f01a811560bfde7523211c35f71d7&vid=d779d50cfe3f01a811560bfde7523211c35f71d7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty