Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-[4-(2-Benzimidazolyl)phenyl]maleimide ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd

B1149-50MG B1149-100MG
TCB1149-050MGEA 183 USD InStock
TCB1149-050MG TCB1149-100MG
N-[4-(2-Benzimidazolyl)phenyl]maleimide ≥98.0% (by HPLC, titration analysis)
N-[4-(2-Benzimidazolyl)phenyl]maleimide
CAS Number: 27030-97-3 MDL Number: MFCD00042767 Molecular Formula: C17H11N3O2 Molecular Weight: 289.29 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 247


Formula: C₁₇H₁₁N₃O₂
Melting Pt: 247 °C
MDL Number: MFCD00042767
CAS Number: 27030-97-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8311bec48c2cd2a4a61ca00322201d5ce4cbaed5&vid=8311bec48c2cd2a4a61ca00322201d5ce4cbaed5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty