Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-Biphenyldisulfonyl Chloride ≥97.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

B1165-5G B1165-25G
TCB1165-005GEA 92 USD InStock
TCB1165-005G TCB1165-025G
4,4'-Biphenyldisulfonyl Chloride ≥97.0% (by titrimetric analysis)
4,4'-Biphenyldisulfonyl Chloride
CAS Number: 3406-84-6
MDL Number: MFCD00007447
Molecular Formula: C12H8Cl2O4S2
Molecular Weight: 351.21
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 207

Product Categories: Materials Chemistry, Biphenyls (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Biphenyldisulfonyl Dichloride, 4,4'-Diphenyldisulfonyl Chloride


Formula: C₁₂H₈Cl₂O₄S₂
Melting Pt: 207 °C
MDL Number: MFCD00007447
CAS Number: 3406-84-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=146445cdf237da5e9a072fcb3941dbae42bc7427&vid=146445cdf237da5e9a072fcb3941dbae42bc7427&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty