Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyldimethylstearylammonium chloride ≥98.0%

Supplier: Decon Laboratories Ltd

B1297-25G B1297-100G
TCB1297-025GEA 80.27 USD InStock
TCB1297-025G TCB1297-100G
Benzyldimethylstearylammonium chloride ≥98.0%
Benzyldimethylstearylammonium chloride
CAS Number: 122-19-0
MDL Number: MFCD00011776
Molecular Formula: C27H50ClN
Molecular Weight: 424.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Product Categories: Materials Chemistry, Cationic Surfactants
Other Product Names / Synonyms: Octadecyldimethylbenzylammonium Chloride, Benzyldimethyloctadecylammonium Chloride


Formula: C₂₇H₅₀ClN
MDL Number: MFCD00011776
CAS Number: 122-19-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47022f32f49aab792e579e79968e4791f51cdbf7&vid=47022f32f49aab792e579e79968e4791f51cdbf7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty