Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Bis(4-chloroformyloxyphenyl)propane ≥97.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

B1335-1G B1335-5G
TCB1335-001GEA 22 USD InStock
TCB1335-001G TCB1335-005G
2,2-Bis(4-chloroformyloxyphenyl)propane ≥97.0% (by GC, titration analysis)
2,2-Bis(4-chloroformyloxyphenyl)propane
CAS Number: 2024-88-6
MDL Number: MFCD00060109
Molecular Formula: C17H14Cl2O4
Molecular Weight: 353.20
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 216
Melting point (°C): 94

Product Categories: Materials Chemistry, Bisphenol A type Compounds (for High-Performance Polymer Research)
Other Product Names / Synonyms: Bisphenol A Bis(chloroformate), 4,4'-Isopropylidenediphenol Bis(chloroformate)


Formula: C₁₇H₁₄Cl₂O₄
Boiling Pt: 216 °C
Melting Pt: 94 °C
MDL Number: MFCD00060109
CAS Number: 2024-88-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96e3cbbfcef0cee38612194f23a33c5d1a592d96&vid=96e3cbbfcef0cee38612194f23a33c5d1a592d96&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty