Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-tert-Butyl-2-methoxyethylamine ≥98.0%

Supplier: Decon Laboratories Ltd

B1430-1G B1430-5G
TCB1430-001GEA 83.7 USD InStock
TCB1430-001G TCB1430-5G
N-tert-Butyl-2-methoxyethylamine ≥98.0%
N-tert-Butyl-2-methoxyethylamine
MDL Number: MFCD00145008
Molecular Formula: C7H17NO
Molecular Weight: 131.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Specific Gravity (20/20): 0.82

Other Product Names / Synonyms: N-(2-Methoxyethyl)-tert-butylamine


https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba210b6d3277aa7aea37ef8cea76bd5d565fd64a&vid=ba210b6d3277aa7aea37ef8cea76bd5d565fd64a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty