Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Benzyloxyphenylacetyl chloride ≥96.0%

Supplier: Decon Laboratories Ltd

B1780-5G B1780-25G
TCB1780-005GEA 171 USD InStock
TCB1780-005G TCB1780-25G
4-Benzyloxyphenylacetyl chloride ≥96.0%
4-Benzyloxyphenylacetyl chloride
CAS Number: 39188-62-0
MDL Number: MFCD00674192
Molecular Formula: C15H13ClO2
Molecular Weight: 260.72
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 78


Formula: C₁₅H₁₃ClO₂
MW: 260.72 g/mol
Melting Pt: 78 °C
MDL Number: MFCD00674192
CAS Number: 39188-62-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f6e66503608a5257e9ae7e5c130f141c26ade19&vid=3f6e66503608a5257e9ae7e5c130f141c26ade19&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty