Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(2-ethylhexanoato)hydroxyaluminum ≥96.0%

Supplier: Decon Laboratories Ltd

B1895-25G B1895-250G
TCB1895-025GEA 17 USD InStock
TCB1895-025G TCB1895-250G
Bis(2-ethylhexanoato)hydroxyaluminum ≥96.0%
Bis(2-ethylhexanoato)hydroxyaluminum
CAS Number: 30745-55-2
MDL Number: MFCD00041940
Molecular Formula: C16H31AlO5
Molecular Weight: 330.40
Purity/Analysis Method: >96.0% (T)
Form: Crystal

Other Product Names / Synonyms: Hydroxyaluminum Bis(2-ethylhexanoate)


Formula: C₁₆H₃₁AlO₅
MDL Number: MFCD00041940
CAS Number: 30745-55-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02b3c360325bd2aba408990b3c3808034145cfaf&vid=02b3c360325bd2aba408990b3c3808034145cfaf&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty