Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2-ethylbutane ≥97.0% stabilised for synthesis

Supplier: Decon Laboratories Ltd

B1905-25G B1905-5G
TCB1905-025GEA 196.7 USD InStock
TCB1905-025G TCB1905-5G
1-Bromo-2-ethylbutane ≥97.0% stabilised for synthesis
1-Bromo-2-ethylbutane
(stabilized with Copper chip)
CAS Number: 3814-34-4
MDL Number: MFCD00000219
Molecular Formula: C6H13Br
Molecular Weight: 165.07
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Flash Point (°C): 36
Specific Gravity (20/20): 1.19

Other Product Names / Synonyms: 3-(Bromomethyl)pentane, 2-Ethylbutyl Bromide


Formula: C₆H₁₃Br
MW: 165.07 g/mol
Boiling Pt: 145 °C
Density: 1.19 (20 °C)
Flash Pt: 36 °C
MDL Number: MFCD00000219
CAS Number: 3814-34-4
EINECS: 223-302-6
UN: 1993
ADR: 3 III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7ea9a3c9b3bd08d7fdc1019be3e84d0a83136aa&vid=a7ea9a3c9b3bd08d7fdc1019be3e84d0a83136aa&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty