Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Bis(2,3,4,5,6-pentabromophenyl)ethane ≥96.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

B2795-500G B2795-25G
TCB2795-500GEA 131 USD InStock
TCB2795-500G TCB2795-25G
1,2-Bis(2,3,4,5,6-pentabromophenyl)ethane ≥96.0% (by titrimetric analysis)
1,2-Bis(2,3,4,5,6-pentabromophenyl)ethane
CAS Number: 84852-53-9
Molecular Formula: C14H4Br10
Molecular Weight: 971.23
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 352

Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: 2,2',3,3',4,4',5,5',6,6'-Decabromobibenzyl


Formula: C₁₄H₄Br₁₀
Melting Pt: 352 °C
MDL Number: MFCD06407713
CAS Number: 84852-53-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=430974cda0634bde1794a6c160eabbb43463dab2&vid=430974cda0634bde1794a6c160eabbb43463dab2&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty