Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzyl-D-aspartate ≥98.0%

Supplier: Decon Laboratories Ltd

B2993-1G B2993-5G
TCB2993-1GEA 66 USD InStock
TCB2993-1G TCB2993-5G
1-Benzyl-D-aspartate ≥98.0%
1-Benzyl-D-aspartate
CAS Number: 79337-40-9
Molecular Formula: C11H13NO4
Molecular Weight: 223.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 176
Specific rotation [a]20/D: 14 deg (C=1, 0.1mol/L HCl)

Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: D-Aspartic Acid 1-Benzyl Ester, H-D-Asp-OBzl


Formula: C₁₁H₁₃NO₄
Melting Pt: 176 °C
MDL Number: MFCD00037289
CAS Number: 79337-40-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df13b2e2e83a44c7a09e3e9227c5f68cdb9e2348&vid=df13b2e2e83a44c7a09e3e9227c5f68cdb9e2348&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty