Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloroacetamide ≥98.0%

Supplier: Decon Laboratories Ltd
Danger

C0086-500G C0086-25G
TCC0086-500GEA 55 USD InStock
TCC0086-500G TCC0086-025G
Chloroacetamide ≥98.0%
Chloroacetamide
CAS Number: 79-07-2
MDL Number: MFCD00008027
Molecular Formula: C2H4ClNO
Molecular Weight: 93.51
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 225
Melting point (°C): 120
Flash Point (°C): 170


Formula: C₂H₄ClNO
Boiling Pt: 225 °C
Melting Pt: 120 °C
Density: 1.6 g/cm³ (20 °C)
Flash Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00008027
CAS Number: 79-07-2
EINECS: 201-174-2
UN: 2811
ADR: 6.1,III
Merck Index: 13,02127
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de851f0e55aa7afcea4b57628c6377716e9d8dfb&vid=de851f0e55aa7afcea4b57628c6377716e9d8dfb&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty