Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-o-anisidine ≥98.0% for synthesis

Supplier: Decon Laboratories Ltd

C0118-25G C0118-500G
TCC0118-025GEA 37 USD InStock
TCC0118-025G TCC0118-500G
5-Chloro-o-anisidine ≥98.0% for synthesis
5-Chloro-o-anisidine
CAS Number: 95-03-4
MDL Number: MFCD00007777
Molecular Formula: C7H8ClNO
Molecular Weight: 157.60
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 82

Other Product Names / Synonyms: 5-Chloro-o-anisidine, 4-Chloro-2-aminoanisole, 2-Amino-4-chloroanisole


Formula: C₇H₈ClNO
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 82 °C
MDL Number: MFCD00007777
CAS Number: 95-03-4
EINECS: 202-385-2
UN: 2233
ADR: 6.1,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53f7b1ddb5cec9396fc92167ed44fdbb41586dec&vid=53f7b1ddb5cec9396fc92167ed44fdbb41586dec&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty