Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Chloromethoxy)ethane ≥96.0% for synthesis

Supplier: Decon Laboratories Ltd

C0201-500G C0201-25G C0201-100G
TCC0201-500GEA 780 USD InStock
TCC0201-500G TCC0201-025G TCC0201-100G
(Chloromethoxy)ethane ≥96.0% for synthesis
(Chloromethoxy)ethane
CAS Number: 3188-13-4
MDL Number: MFCD00000887
Molecular Formula: C3H7ClO
Molecular Weight: 94.54
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 7
Specific Gravity (20/20): 1.02


Formula: ClCH₂OCH₂CH₃
MW: 94.54 g/mol
Boiling Pt: 80 °C
Melting Pt: –98 °C
Density: 1.02 (20 °C)
Flash Pt: 7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000887
CAS Number: 3188-13-4
EINECS: 221-687-5
UN: 2354
ADR: 3 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cb737e98551ae005790ae1c446287f4ead64b4b&vid=5cb737e98551ae005790ae1c446287f4ead64b4b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty