Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Picrotoxin (picrotoxinin + picrotin) ≥94.0% (by HPLC)

Supplier: Decon Laboratories Ltd

C0375-1G C0375-5G
TCC0375-001GEA 120.8 USD InStock
TCC0375-001G TCC0375-005G
Picrotoxin (picrotoxinin + picrotin) ≥94.0% (by HPLC)
Picrotoxin (picrotoxinin + picrotin)
CAS Number: 124-87-8
MDL Number: MFCD00074824
Molecular Formula: C30H34O13
Molecular Weight: 602.59
Purity/Analysis Method: >94.0% (HPLC)
Form: Crystal
Melting point (°C): 200
Specific rotation [a]20/D: -34 deg (C=4, EtOH)

Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Sesquiterpenes
Other Product Names / Synonyms: Cocculin


Formula: C₃₀H₃₄O₁₃
Melting Pt: 200 °C
MDL Number: MFCD00074824
CAS Number: 124-87-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f25b7d41c812cfc5d8f631232181d4efb7e4833&vid=9f25b7d41c812cfc5d8f631232181d4efb7e4833&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >94.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty