C0503-500ML
C0503-25ML
TCC0503-500MLEA
87.16
USD
InStock
TCC0503-500ML
TCC0503-025ML
1,5-Cyclooctadiene ≥98.0% stabilised for synthesis
1,5-Cyclooctadiene
[stabilized with Octadecyl 3-(3',5'-Di-tert-butyl-4'-hydroxyphenyl)propionate]
CAS Number: 111-78-4
MDL Number: MFCD00001752
Molecular Formula: C8H12
Molecular Weight: 108.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Melting point (°C): -69
Flash Point (°C): 35
Specific Gravity (20/20): 0.88
Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: COD