Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenyl-3-chloropropionate ≥95.0%

Supplier: Decon Laboratories Ltd

C0850-500ML C0850-25ML
TCC0850-500MLEA 369.3 USD InStock
TCC0850-500ML TCC0850-025ML
Phenyl-3-chloropropionate ≥95.0%
Phenyl-3-chloropropionate
CAS Number: 24552-27-0
MDL Number: MFCD00045298
Molecular Formula: C9H9ClO2
Molecular Weight: 184.62
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 157
Specific Gravity (20/20): 1.20

Other Product Names / Synonyms: 3-Chloropropionic Acid Phenyl Ester


Formula: C₉H₉ClO₂
Boiling Pt: 157 °C
Density: 1.20 (20 °C)
MDL Number: MFCD00045298
CAS Number: 24552-27-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46652cdc58f3f11d11f0ecd6e59ad18bc0a0b455&vid=46652cdc58f3f11d11f0ecd6e59ad18bc0a0b455&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty