Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(4-Chlorobutyl)hydantoin ≥97.0%

Supplier: Decon Laboratories Ltd

C0902-5G C0902-25G
TCC0902-5GEA 107.8 USD InStock
TCC0902-5G TCC0902-025G
5-(4-Chlorobutyl)hydantoin ≥97.0%
5-(4-Chlorobutyl)hydantoin
CAS Number: 40126-55-4
MDL Number: MFCD00059722
Molecular Formula: C7H11ClN2O2
Molecular Weight: 190.63
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 131


Formula: C₇H₁₁ClN₂O₂
MW: 190.63 g/mol
Melting Pt: 131 °C
MDL Number: MFCD00059722
CAS Number: 40126-55-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed706c5aeda8cb4396fb96f09512e1d1a01f18f8&vid=ed706c5aeda8cb4396fb96f09512e1d1a01f18f8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty