Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

sec-Butyl crotonate ≥97.0%

Supplier: Decon Laboratories Ltd

C1184-25ML C1184-250ML
TCC1184-025MLEA 80 USD InStock
TCC1184-025ML TCC1184-250ML
sec-Butyl crotonate ≥97.0%
sec-Butyl crotonate
CAS Number: 10371-45-6
MDL Number: MFCD00059054
Molecular Formula: C8H14O2
Molecular Weight: 142.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 166
Specific Gravity (20/20): 0.89

Other Product Names / Synonyms: Crotonic Acid sec-Butyl Ester


Formula: C₈H₁₄O₂
Boiling Pt: 166 °C
Density: 0.89 (20 °C)
MDL Number: MFCD00059054
CAS Number: 10371-45-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=688e6756c0e537147cab2f7d06094e7b55d2b2a8&vid=688e6756c0e537147cab2f7d06094e7b55d2b2a8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty