Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-2-Chlorobutyric acid ≥98.0%

Supplier: Decon Laboratories Ltd

C1377-5G C1377-1G
TCC1377-005GEA 552 USD InStock
TCC1377-005G TCC1377-001G
(S)-(-)-2-Chlorobutyric acid ≥98.0%
(S)-(-)-2-Chlorobutyric acid
CAS Number: 32653-32-0
MDL Number: MFCD00067099
Molecular Formula: C4H7ClO2
Molecular Weight: 122.55
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 94
Specific Gravity (20/20): 1.19
Specific rotation [a]20/D: -16 deg (neat)


Formula: C₄H₇ClO₂
Boiling Pt: 94 °C
Density: 1.19 (20 °C)
MDL Number: MFCD00067099
CAS Number: 32653-32-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf858c702ab1aa8e2026e907f855aed3b0d82395&vid=cf858c702ab1aa8e2026e907f855aed3b0d82395&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty