Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Carbobenzoxy-L-serine methyl ester ≥98.0%

Supplier: Decon Laboratories Ltd

C1403-5G C1403-25G
TCC1403-005GEA 92 USD InStock
TCC1403-005G TCC1403-25G
N-Carbobenzoxy-L-serine methyl ester ≥98.0%
N-Carbobenzoxy-L-serine methyl ester
CAS Number: 1676-81-9
MDL Number: MFCD00077039
Molecular Formula: C12H15NO5
Molecular Weight: 253.25
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 37
Flash Point (°C): 113
Specific rotation [a]20/D: -15 deg (C=1, MeOH)
Storage Temperature: 0-10°C

Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-L-serine Methyl Ester, Z-Ser-OMe


Formula: C₁₂H₁₅NO₅
Melting Pt: 37 °C
Flash Pt: 113 °C
MDL Number: MFCD00077039
CAS Number: 1676-81-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=424bff13b3fa51683ef2911316b8aaaa8c2729a8&vid=424bff13b3fa51683ef2911316b8aaaa8c2729a8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (N)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty