Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-1,4-Diaminocyclohexane ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

C1426-25G C1426-5G
TCC1426-025GEA 41.9 USD InStock
TCC1426-025G TCC1426-005G
trans-1,4-Diaminocyclohexane ≥98.0% (by GC, titration analysis)
trans-1,4-Diaminocyclohexane
CAS Number: 2615-25-0
MDL Number: MFCD00075174
Molecular Formula: C6H14N2
Molecular Weight: 114.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 88
Melting point (°C): 71
Flash Point (°C): 71

Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: trans-1,4-Diaminocyclohexane, trans-Hexahydro-1,4-phenylenediamine


Formula: C₆H₁₄N₂
MDL Number: MFCD00075174
CAS Number: 2615-25-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e487645772c0893245adb3267c04446333806f1b&vid=e487645772c0893245adb3267c04446333806f1b&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty