Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Clarithromycin ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

C2220-5G C2220-1G C2220-25G
TCC2220-5GEA 309.02 USD InStock
TCC2220-5G TCC2220-1G TCC2220-25G
Clarithromycin ≥98.0% (by titrimetric analysis)
Clarithromycin
CAS Number: 81103-11-9
MDL Number: MFCD00865140
Molecular Formula: C38H69NO13
Molecular Weight: 747.96
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 225
Specific rotation [a]20/D: -92 deg (C=1, CHCl3)


Formula: C₃₈H₆₉NO₁₃
Melting Pt: 225 °C
MDL Number: MFCD00865140
CAS Number: 81103-11-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed6def2a2843febaba49370e913a45311ea242ba&vid=ed6def2a2843febaba49370e913a45311ea242ba&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty