Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ceftazidime (contains ca. 10% Na2CO3)

Supplier: Decon Laboratories Ltd

C2225-25G C2225-5G
TCC2225-25GEA 340 USD InStock
TCC2225-25G TCC2225-5G
Ceftazidime (contains ca. 10% Na2CO3)
Ceftazidime
CAS Number: 72558-82-8
MDL Number: MFCD00072034
Molecular Formula: C22H22N6O7S2
Molecular Weight: 546.57
Form: Crystal
Color: White

Other Product Names / Synonyms: CAZ


Formula: C₂₂H₂₂N₆O₇S₂
MDL Number: MFCD00072034
CAS Number: 72558-82-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dcb7b393196a69900a356372d185263a4a1a0e7&vid=4dcb7b393196a69900a356372d185263a4a1a0e7&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty