Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cephalexin monohydrate ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd

C2248-25G C2248-5G
TCC2248-25GEA 330 USD InStock
TCC2248-25G TCC2248-5G
Cephalexin monohydrate ≥98.0% (by HPLC, titration analysis)
Cephalexin monohydrate
CAS Number: 23325-78-2
MDL Number: MFCD00167148
Molecular Formula: C16H17N3O4S
Molecular Weight: 347.39
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: 154 deg (C=0.5, H2O)

Product Categories: Bioscience, Antibiotic Ingredients
Other Product Names / Synonyms: Cefalexin


Formula: C₁₆H₁₇N₃O₄S
MDL Number: MFCD00167148
CAS Number: 23325-78-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=343ebd38715d4da81c561f15df58709264103e68&vid=343ebd38715d4da81c561f15df58709264103e68&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty