Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Curcumin ≥97.0%

Supplier: Decon Laboratories Ltd

C2302-25G C2302-5G
TCC2302-25GEA 186 USD InStock
TCC2302-25G TCC2302-5G
Curcumin ≥97.0%
Curcumin
CAS Number: 458-37-7
MDL Number: MFCD00008365
Molecular Formula: C21H20O6
Molecular Weight: 368.39
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 186
Storage Temperature: <0°C

Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Immune System, Inflammation, Allergy Research Reagents, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Antioxidants, Anti-inflamm
Other Product Names / Synonyms: (E,E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione


Formula: C₂₁H₂₀O₆
MW: 368.39 g/mol
Melting Pt: 186 °C
MDL Number: MFCD00008365
CAS Number: 458-37-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c75cd166f0e6ee6434fad408ed12ac3c413e7461&vid=c75cd166f0e6ee6434fad408ed12ac3c413e7461&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty