Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Proflavine hemisulfate hydrate ≥97.0% (by total nitrogen basis)

Supplier: Decon Laboratories Ltd

D0073-5G D0073-25G
TCD0073-005GEA 44 USD InStock
TCD0073-005G TCD0073-025G
Proflavine hemisulfate hydrate ≥97.0% (by total nitrogen basis)
Proflavine hemisulfate hydrate
CAS Number: 1811-28-5
MDL Number: MFCD00013150
Molecular Formula: C13H11N3
Molecular Weight: 516.58
Purity/Analysis Method: >97.0% (N)
Form: Crystal

Product Categories: Bioscience, Stains & Dyes, Fluorescent Stains, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 3,6-Diaminoacridinium Monohydrogen Sulfate


Formula: C₁₃H₁₁N₃
MDL Number: MFCD00013150
CAS Number: 1811-28-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6733de2fa794bffb0277c5151db3b5c35ef06d88&vid=6733de2fa794bffb0277c5151db3b5c35ef06d88&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty