Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dibromo-2-propanol ≥95.0% stabilised for synthesis

Supplier: Decon Laboratories Ltd

D0187-500ML D0187-25ML D0187-100ML
TCD0187-500MLEA 1114 USD InStock
TCD0187-500ML TCD0187-025ML TCD0187-100ML
1,3-Dibromo-2-propanol ≥95.0% stabilised for synthesis
1,3-Dibromo-2-propanol
(stabilized with Copper chip)
CAS Number: 96-21-9
MDL Number: MFCD00000216
Molecular Formula: C3H6Br2O
Molecular Weight: 217.89
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 215
Flash Point (°C): 46
Specific Gravity (20/20): 2.13
Storage Temperature: 0-10°C

Other Product Names / Synonyms: alpha,alpha'-Dibromohydrin, Glycerol alpha,alpha'-Dibromohydrin


Formula: BrH₂CCH(OH)CH₂Br
MW: 217.89 g/mol
Boiling Pt: 89
Density: 2.13 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00000216
CAS Number: 96-21-9
EINECS: 202-489-8
UN: 1992
ADR: 3 III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5afc7a2191b3616a7e9f9dc3d94e620bdb46bcee&vid=5afc7a2191b3616a7e9f9dc3d94e620bdb46bcee&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty