Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichloramine T ≥96.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

D0318-25G D0318-500G
TCD0318-025GEA 96.67 USD InStock
TCD0318-025G TCD0318-500G
Dichloramine T ≥96.0% (by titrimetric analysis)
Dichloramine T
CAS Number: 473-34-7
MDL Number: MFCD00058929
Molecular Formula: C7H7Cl2NO2S
Molecular Weight: 240.10
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 79

Product Categories: Synthetic Organic Chemistry, Oxidation, Halogenation
Other Product Names / Synonyms: N,N-Dichloro-p-toluenesulfonamide, p-Toluenesulfondichloramide


Formula: C₇H₇Cl₂NO₂S
Melting Pt: 79 °C
MDL Number: MFCD00058929
CAS Number: 473-34-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a66e59164889c96d62c840fb2b493ca86100546&vid=0a66e59164889c96d62c840fb2b493ca86100546&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty