Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Guanylurea phosphate ≥98.0%

Supplier: Decon Laboratories Ltd

D0432-500G D0432-25G
TCD0432-500GEA 214 USD InStock
TCD0432-500G TCD0432-025G
Guanylurea phosphate ≥98.0%
Guanylurea phosphate
CAS Number: 36897-89-9
MDL Number: MFCD00065097
Molecular Formula: C2H6N4O
Molecular Weight: 200.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 184

Other Product Names / Synonyms: Dicyandiamidine Phosphate


Formula: C₂H₆N₄O·H₃PO₄
MW: 200.09 g/mol
Melting Pt: 184 °C
MDL Number: MFCD00065097
CAS Number: 36897-89-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7d5a9c2079441977535f0c1e08219c7c1f1d096&vid=b7d5a9c2079441977535f0c1e08219c7c1f1d096&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty