Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Diethylamino)-1-propanol ≥95.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

D0472-25ML D0472-500ML
TCD0472-025MLEA 103 USD InStock
TCD0472-025ML TCD0472-500ML
3-(Diethylamino)-1-propanol ≥95.0% (by GC, titration analysis)
3-(Diethylamino)-1-propanol
CAS Number: 622-93-5
MDL Number: MFCD00002947
Molecular Formula: C7H17NO
Molecular Weight: 131.22
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 189
Flash Point (°C): 65
Specific Gravity (20/20): 0.88


Formula: C₇H₁₇NO
Boiling Pt: 189 °C
Density: 0.88 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00002947
CAS Number: 622-93-5
EINECS: 210-759-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f4fab5003e41fc3f970c204105870963e5007a9&vid=2f4fab5003e41fc3f970c204105870963e5007a9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty