Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-N,N-diethylaniline ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd
Danger

D0518-500G D0518-25G
TCD0518-500GEA 1708.4 USD InStock
TCD0518-500G TCD0518-025G
4-Amino-N,N-diethylaniline ≥98.0% (by GC, titration analysis)
4-Amino-N,N-diethylaniline
CAS Number: 93-05-0
MDL Number: MFCD00007861
Molecular Formula: C10H16N2
Molecular Weight: 164.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 262
Flash Point (°C): 139
Freezing point (°C): 18
Storage Temperature: <0°C

Other Product Names / Synonyms: 4-Amino-N,N-diethylaniline


Formula: C₁₀H₁₆N₂
Boiling Pt: 262 °C
Melting Pt: 23 °C
Density: 0.988 g/cm³ (20 °C)
Flash Pt: 139 °C
MDL Number: MFCD00007861
CAS Number: 93-05-0
EINECS: 202-214-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa98be602cc1e907e00f57ed7461f5e7b2e3adf6&vid=aa98be602cc1e907e00f57ed7461f5e7b2e3adf6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty