Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl sulfite ≥98.0%

Supplier: Decon Laboratories Ltd

D0528-500G D0528-25G
TCD0528-500GEA 417 USD InStock
TCD0528-500G TCD0528-025G
Diethyl sulfite ≥98.0%
Diethyl sulphite
CAS Number: 623-81-4
MDL Number: MFCD00009098
Molecular Formula: C4H10O3S
Molecular Weight: 138.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 157
Flash Point (°C): 49
Specific Gravity (20/20): 1.09


Formula: C₄H₁₀O₃S
Boiling Pt: 157 °C
Density: 1.09 (20 °C)
Flash Pt: 49 °C
MDL Number: MFCD00009098
CAS Number: 623-81-4
EINECS: 210-815-5
UN: 1993
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=591a5cf64b75ee8f3a37da291180944e576e2c15&vid=591a5cf64b75ee8f3a37da291180944e576e2c15&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty