Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diphenyl phosphite ≥75.0% (contains ca. 5% Phenol)

Supplier: Decon Laboratories Ltd

D0907-25G D0907-500G
TCD0907-025GEA 17.12 USD InStock
TCD0907-025G TCD0907-500G
Diphenyl phosphite ≥75.0% (contains ca. 5% Phenol)
Diphenyl phosphite
CAS Number: 4712-55-4
MDL Number: MFCD00044497
Molecular Formula: C12H11O3P
Molecular Weight: 234.19
Purity/Analysis Method: >75.0% (HPLC)
Form: Clear Liquid
Boiling point (°C): 148
Melting point (°C): 12
Flash Point (°C): 186
Specific Gravity (20/20): 1.22

Product Categories: Synthetic Organic Chemistry, Dialkyl Phosphites (Multi-component Reaction)
Other Product Names / Synonyms: Diphenyl Phosphonate, Phosphonic Acid Diphenyl Ester, Phosphorous Acid Diphenyl Ester


Formula: C₁₂H₁₁O₃P
Boiling Pt: 148 °C
Melting Pt: 12 °C
Density: 1.22 (20 °C)
Flash Pt: 186 °C
MDL Number: MFCD00044497
CAS Number: 4712-55-4
UN: 3265
ADR: 8,I
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f48535dff456396074a2370dd67837203e6614a&vid=0f48535dff456396074a2370dd67837203e6614a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >75.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty