Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Dodecen-1-yl)succinic anhydride (mixture of cis- and trans-isomeres) ≥95.0%

Supplier: Decon Laboratories Ltd

D0976-25G D0976-500G
TCD0976-025GEA 39 USD InStock
TCD0976-025G TCD0976-500G
(2-Dodecen-1-yl)succinic anhydride (mixture of cis- and trans-isomeres) ≥95.0%
(2-Dodecen-1-yl)succinic anhydride (mixture of cis- and trans-isomeres)
CAS Number: 19780-11-1
MDL Number: MFCD00005528
Molecular Formula: C16H26O3
Molecular Weight: 266.38
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Flash Point (°C): 185
Freezing point (°C): 32


Formula: C₁₆H₂₆O₃
MW: 266.375 g/mol
Flash Pt: 185 °C
MDL Number: MFCD00005528
CAS Number: 19780-11-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d50b291a6a513db6dcc78c6a7e3071e190af2d9d&vid=d50b291a6a513db6dcc78c6a7e3071e190af2d9d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty